Assay Details
Gene Symbol: CCDC188 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028827968.1 | XP_028683801.1 | 1118 | 78 | ||
XM_028827969.1 | XP_028683802.1 | 1118 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
