Assay Details
Gene Symbol: TIRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028832728.1 | XP_028688561.1 | 121 | 107 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028832728.1 transcripts.
Set Membership: |
![]() ![]() |
