Assay Details
Gene Symbol: LOC716404 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028836627.1 | XP_028692460.1 | 7602 | 103 | ||
XM_028836628.1 | XP_028692461.1 | 7119 | 103 |

More Information
Set Membership: |
![]() ![]() |
