Assay Details
Gene Symbol: COMMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015112466.2 | XP_014967952.2 | 205 | 109 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015112466.2 transcripts.
Set Membership: |
![]() ![]() |
