Assay Details
Gene Symbol: HOXB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028836403.1 | XP_028692236.1 | 415 | 88 | ||
XM_028836404.1 | XP_028692237.1 | 415 | 88 | |||
XM_028836405.1 | XP_028692238.1 | 415 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028836403.1, XM_028836404.1, XM_028836405.1 transcripts.
Set Membership: |
![]() ![]() |
