Assay Details
Gene Symbol: P2RY6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001115246.4 | XP_001115246.2 | 664 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001115246.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
