Assay Details
Gene Symbol: CEBPZOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028831759.1 | XP_028687592.1 | 281 | 118 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028831759.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
