Assay Details
Gene Symbol: FEZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015134799.2 | XP_014990285.1 | 1620 | 54 | ||
XM_015134800.2 | XP_014990286.1 | 1618 | 54 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015134799.2, XM_015134800.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
