Assay Details
Gene Symbol: LOC100430319 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015151039.2 | XP_015006525.2 | 814 | 100 | ||
XM_015151040.2 | XP_015006526.2 | 814 | 100 | |||
XM_015151041.2 | XP_015006527.2 | 814 | 100 |
