Assay Details
Gene Symbol: NDUFA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028839290.1 | XP_028695123.1 | 159 | 109 | ||
XM_028839291.1 | XP_028695124.1 | 159 | 109 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028839290.1, XM_028839291.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
