Assay Details
Gene Symbol: ERRFI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261681.1 | NP_001248610.1 | 1 - 2 | 77 | 99 | |
GenBank mRNA | JU335057.1 | - | 1 - 2 | 124 | 99 | |
JV047417.1 | - | 1 - 2 | 77 | 99 | ||
JV619708.1 | - | 1 - 2 | 161 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001261681.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
