Assay Details
Gene Symbol: PNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193551.1 | NP_001180480.1 | 3 - 4 | 413 | 86 | |
GenBank mRNA | CN646333.1 | - | 3 - 4 | 413 | 86 | |
JU323241.1 | - | 3 - 4 | 447 | 86 | ||
JU475742.1 | - | 3 - 4 | 367 | 86 | ||
JV045361.1 | - | 3 - 4 | 366 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
