Assay Details
Gene Symbol: Pten | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031606.1 | NP_113794.1 | 1 - 2 | 77 | 114 | |
GenBank mRNA | AF017185.1 | - | 1 - 2 | 77 | 114 | |
AF455569.1 | - | 1 - 2 | 77 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
