Assay Details
Gene Symbol: Cacnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012828.2 | NP_036960.1 | 1 - 2 | 137 | 57 | |
GenBank mRNA | CB739910.1 | - | 1 - 2 | 263 | 57 | |
M88751.1 | - | 1 - 2 | 141 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
