Assay Details
Gene Symbol: Cacnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012828.2 | NP_036960.1 | 2 - 3 | 261 | 85 | |
XM_006257329.2 | XP_006257391.1 | 2 - 3 | 333 | 85 | ||
XM_006257330.2 | XP_006257392.1 | 2 - 3 | 742 | 85 | ||
GenBank mRNA | CB739910.1 | - | 2 - 3 | 387 | 85 | |
M88751.1 | - | 2 - 3 | 265 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
