Assay Details
Gene Symbol: Esrra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006230755.1 | XP_006230817.1 | 1 - 2 | 221 | 154 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
