Assay Details
Gene Symbol: Gabrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246125.2 | XP_006246187.1 | 4 - 5 | 1111 | 87 | |
GenBank mRNA | AY574252.1 | - | 4 - 5 | 548 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
