Assay Details
Gene Symbol: Pparg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013124.3 | NP_037256.1 | 1 - 2 | 165 | 63 | |
GenBank mRNA | AB019561.1 | - | 1 - 2 | 79 | 63 | |
AF156666.1 | - | 1 - 2 | 127 | 63 | ||
Y12882.2 | - | 1 - 2 | 128 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
