Assay Details
Gene Symbol: Kitlg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021843.4 | NP_068615.1 | 7 - 8 | 882 | 72 | |
NM_021844.2 | NP_068616.1 | 6 - 7 | 798 | 72 | ||
XM_008765339.1 | XP_008763561.1 | 7 - 8 | 926 | 72 | ||
XM_008765340.1 | XP_008763562.1 | 6 - 7 | 842 | 72 | ||
GenBank mRNA | AB105879.1 | - | 6 - 7 | 798 | 72 | |
AF071204.1 | - | 7 - 8 | 717 | 72 | ||
AF071205.1 | - | 6 - 7 | 634 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
ovarian follicle development
neural crest cell migration
positive regulation of myeloid leukocyte differentiation
cell adhesion
positive regulation of cell population proliferation
male gonad development
negative regulation of mast cell apoptotic process
embryonic hemopoiesis
ectopic germ cell programmed cell death
negative regulation of apoptotic process
positive regulation of MAP kinase activity
positive regulation of melanocyte differentiation
positive regulation of DNA replication
positive regulation of Ras protein signal transduction
positive regulation of peptidyl-tyrosine phosphorylation
positive regulation of mast cell proliferation
extrinsic apoptotic signaling pathway in absence of ligand
cytokine activity
stem cell factor receptor binding
growth factor activity
ovarian follicle development
neural crest cell migration
positive regulation of myeloid leukocyte differentiation
cell adhesion
positive regulation of cell population proliferation
male gonad development
negative regulation of mast cell apoptotic process
embryonic hemopoiesis
ectopic germ cell programmed cell death
negative regulation of apoptotic process
positive regulation of MAP kinase activity
positive regulation of melanocyte differentiation
positive regulation of DNA replication
positive regulation of Ras protein signal transduction
positive regulation of peptidyl-tyrosine phosphorylation
positive regulation of mast cell proliferation
extrinsic apoptotic signaling pathway in absence of ligand
cytokine activity
stem cell factor receptor binding
growth factor activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P054581 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P083349 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P14111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201012 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364396 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500996 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714752 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873494 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755548 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71749 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440917 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149841 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267909 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340596 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970418 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932886 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197101 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531465 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189607 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436471 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82585 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872602 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587591 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58804 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59686 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159683 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146933 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124222 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P054581 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P083349 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P14111 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P083349 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P14111 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P054581 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P14111 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P083349 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P054581 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P14111 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P15811 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 46784 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 57700 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 96597 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 82416 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 40013 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 81984 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10895157 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895156 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895158 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895153 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895164 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895155 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895160 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895161 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895159 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895162 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895163 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10895152 | - | - |
Rat Genome 230 2.0 Array | 1388856_at | - | - |
Rat Genome 230 2.0 Array | 1396214_at | - | - |
Rat Genome 230 2.0 Array | 1388096_at | - | - |
Rat Genome 230 2.0 Array | 1388856_PM_at | - | - |
Rat Genome 230 2.0 Array | 1388096_PM_at | - | - |
Rat Genome 230 2.0 Array | 1396214_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1396214_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388096_at | - | - |
Affymetrix HT Rat Focus Array | 1388856_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1396214_at | - | - |
Affymetrix HT Rat Focus Array | 1388096_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388856_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700005003.rn.1 | TC0700000608.rn.1 | RTA TC |
Rat Expression Set 230 | 1388856_PM_at | - | - |
Rat Expression Set 230 | 1388096_PM_at | - | - |
Rat Expression Set 230 | 1388096_at | - | - |
Rat Expression Set 230 | 1388856_at | - | - |
Rat Expression Set 230 | 1396214_at | - | - |
Rat Expression Set 230 | 1396214_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895152 | - | - |
Rat Genome U34 Set | AF071204_g_at | - | - |
Rat Genome U34 Set | AF071204_at | - | - |
Rat Genome U34 Set | rc_AI102098_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21707765 | - | - |
ABI Rat Genome Survey Microarray | 22371295 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1366908 | - | - |
RatRef-12 Expression BeadChip | ILMN_68344 | - | - |
