Assay Details
Gene Symbol: Tnfsf13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253480.2 | XP_006253542.1 | 4 - 5 | 1222 | 79 | |
XM_006253482.2 | XP_006253544.1 | 3 - 4 | 1125 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
