Assay Details
Gene Symbol: Pcyt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078622.2 | NP_511177.2 | 4 - 5 | 438 | 83 | |
XM_006248444.2 | XP_006248506.1 | 4 - 5 | 591 | 83 | ||
XM_008768694.1 | XP_008766916.1 | 4 - 5 | 789 | 83 | ||
GenBank mRNA | AW919077.1 | - | 4 - 5 | 439 | 83 | |
BC085713.1 | - | 4 - 5 | 436 | 7127059 | 83 | |
L13245.1 | - | 3 - 4 | 335 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
