Assay Details
Gene Symbol: Nap1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053561.2 | NP_446013.2 | 3 - 4 | 268 | 89 | |
XM_006241342.1 | XP_006241404.1 | 3 - 4 | 282 | 89 | ||
XM_006241343.2 | XP_006241405.1 | 3 - 4 | 282 | 89 | ||
GenBank mRNA | AF062594.1 | - | 3 - 4 | 253 | 89 | |
CO574484.1 | - | 284 | 89 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
