Assay Details
Gene Symbol: Nasp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005543.1 | NP_001005543.1 | 6 - 7 | 575 | 114 | |
XM_006238646.2 | XP_006238708.1 | 6 - 7 | 521 | 114 | ||
XM_008763993.1 | XP_008762215.1 | 6 - 7 | 598 | 114 | ||
GenBank mRNA | BC081913.1 | - | 6 - 7 | 575 | 7112076 | 114 |
FQ212041.1 | - | 5 - 6 | 515 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
