Assay Details
Gene Symbol: Ptpn9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013040.1 | NP_001013058.1 | 1 - 2 | 84 | 90 | |
XM_006243106.2 | XP_006243168.1 | 1 - 2 | 676 | 90 | ||
GenBank mRNA | BC082041.1 | - | 1 - 2 | 84 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
