Assay Details
Gene Symbol: Ranbp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008771586.1 | XP_008769808.1 | 12 - 13 | 1867 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
