Assay Details
Gene Symbol: Scn5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160162.1 | NP_001153634.1 | 5 - 6 | 814 | 70 | |
NM_013125.2 | NP_037257.1 | 5 - 6 | 814 | 70 | ||
XM_006244076.2 | XP_006244138.1 | 5 - 6 | 984 | 70 | ||
XM_008766670.1 | XP_008764892.1 | 5 - 6 | 2012 | 70 | ||
XM_008766671.1 | XP_008764893.1 | 5 - 6 | 2012 | 70 | ||
GenBank mRNA | AF353637.1 | - | 4 - 5 | 617 | 70 | |
M27902.1 | - | 5 - 6 | 814 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
