Assay Details
Gene Symbol: Slc13a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031746.1 | NP_113934.1 | 1 - 2 | 159 | 64 | |
XM_006246947.2 | XP_006247009.1 | 1 - 2 | 181 | 64 | ||
GenBank mRNA | AB001321.1 | - | 1 - 2 | 162 | 64 | |
AF058714.1 | - | 1 - 2 | 159 | 64 | ||
U51153.1 | - | 1 - 2 | 151 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
