Assay Details
Gene Symbol: Ttbk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107766.1 | NP_001101236.1 | 2 - 3 | 246 | 81 | |
XM_006234833.2 | XP_006234895.1 | 3 - 4 | 586 | 81 | ||
XM_006234834.2 | XP_006234896.1 | 3 - 4 | 229 | 81 | ||
XM_006234835.2 | XP_006234897.1 | 2 - 3 | 512 | 81 | ||
XM_006234837.2 | XP_006234899.1 | 3 - 4 | 232 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
