Assay Details
Gene Symbol: Cadm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047107.1 | NP_001040572.1 | 2 - 3 | 368 | 55 | |
GenBank mRNA | DQ272745.1 | - | 2 - 3 | 368 | 55 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
