Assay Details
Gene Symbol: Eno3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012949.2 | NP_037081.2 | 2 - 3 | 136 | 79 | |
XM_006246599.2 | XP_006246661.1 | 2 - 3 | 207 | 79 | ||
XM_006246600.2 | XP_006246662.1 | 2 - 3 | 187 | 79 | ||
GenBank mRNA | BC083566.1 | - | 2 - 3 | 123 | 7193009 | 79 |
DY321069.1 | - | 2 - 3 | 136 | 79 | ||
FQ214810.1 | - | 1 - 2 | 114 | 79 | ||
FQ214945.1 | - | 2 - 3 | 154 | 79 | ||
FQ214984.1 | - | 2 - 3 | 146 | 79 | ||
FQ215357.1 | - | 2 - 3 | 158 | 79 | ||
FQ215526.1 | - | 2 - 3 | 146 | 79 | ||
FQ215648.1 | - | 2 - 3 | 146 | 79 | ||
FQ215759.1 | - | 2 - 3 | 158 | 79 | ||
FQ215843.1 | - | 1 - 2 | 113 | 79 | ||
FQ215865.1 | - | 2 - 3 | 146 | 79 | ||
FQ216150.1 | - | 1 - 2 | 114 | 79 | ||
FQ216382.1 | - | 2 - 3 | 146 | 79 | ||
FQ216386.1 | - | 1 - 2 | 123 | 79 | ||
FQ216660.1 | - | 1 - 2 | 113 | 79 | ||
FQ217095.1 | - | 2 - 3 | 146 | 79 | ||
FQ224938.1 | - | 2 - 3 | 146 | 79 | ||
Y00979.1 | - | 1 - 2 | 98 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
