Assay Details
Gene Symbol: Hnrnpab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031330.2 | NP_112620.2 | 2 - 3 | 419 | 58 | |
XM_003750817.3 | XP_003750865.1 | 2 - 3 | 473 | 58 | ||
GenBank mRNA | AF216753.1 | - | 1 - 2 | 220 | 58 | |
AJ238854.1 | - | 2 - 3 | 431 | 58 | ||
AJ238855.1 | - | 2 - 3 | 431 | 58 | ||
BC066664.1 | - | 2 - 3 | 414 | 58 |
Gene Symbol: LOC103689931 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246345.2 | XP_006246407.1 | 2 - 3 | 473 | 58 | |
XM_008767732.1 | XP_008765954.1 | 2 - 3 | 473 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
