Assay Details
Gene Symbol: Arpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106615.2 | NP_001100085.2 | 1 - 2 | 32 | 65 | |
GenBank mRNA | CK478610.1 | - | 1 - 2 | 32 | 65 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001106615.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
