Assay Details
Gene Symbol: Asic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012892.2 | NP_037024.2 | 1 - 2 | 718 | 105 | |
GenBank mRNA | Y14635.1 | - | 1 - 2 | 718 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
