Assay Details
Gene Symbol: Pitpna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017231.1 | NP_058927.1 | 6 - 7 | 566 | 103 | |
GenBank mRNA | BC070945.1 | - | 6 - 7 | 594 | 103 | |
M25758.1 | - | 6 - 7 | 566 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
