Assay Details
Gene Symbol: Nfatc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107805.1 | NP_001101275.1 | 1 - 2 | 213 | 94 | |
XM_006235653.1 | XP_006235715.1 | 1 - 2 | 207 | 94 | ||
XM_008762507.1 | XP_008760729.1 | 1 - 2 | 207 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
