Assay Details
Gene Symbol: Nfe2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031789.2 | NP_113977.1 | 1 - 2 | 113 | 61 | |
XM_006234396.2 | XP_006234458.1 | 1 - 2 | 261 | 61 | ||
GenBank mRNA | AF037350.1 | - | 1 - 2 | 115 | 61 | |
BC061724.1 | - | 1 - 2 | 128 | 5598313 | 61 | |
FQ221441.1 | - | 1 - 2 | 134 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
