Assay Details
Gene Symbol: Cap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053874.1 | NP_446326.1 | 11 - 12 | 1258 | 88 | |
XM_006253753.2 | XP_006253815.1 | 12 - 13 | 1367 | 88 | ||
XM_006253754.2 | XP_006253816.1 | 12 - 13 | 1480 | 88 | ||
GenBank mRNA | U31935.1 | - | 11 - 12 | 1258 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
