Assay Details
Gene Symbol: Cpt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013200.1 | NP_037332.1 | 9 - 10 | 1010 | 82 | |
XM_006242180.2 | XP_006242242.1 | 9 - 10 | 1466 | 82 | ||
GenBank mRNA | BC085761.1 | - | 9 - 10 | 1022 | 7193798 | 82 |
D43623.1 | - | 8 - 9 | 997 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
