Assay Details
Gene Symbol: Msl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014111.1 | NP_001014133.1 | 8 - 9 | 1105 | 88 | |
XM_006256836.2 | XP_006256898.2 | 9 - 10 | 1494 | 88 | ||
XM_006256839.2 | XP_006256901.2 | 9 - 10 | 1491 | 88 | ||
GenBank mRNA | BC086548.1 | - | 8 - 9 | 1105 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
