Assay Details
Gene Symbol: Galns | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047851.1 | NP_001041316.1 | 13 - 14 | 1482 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P114623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173473 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409233 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219399 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653917 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274431 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21478 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136653 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375892 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877717 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30382 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157260 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P114623 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P114623 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P114623 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P405547 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 77710 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 57168 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 4559 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 77996 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10811677 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10811673 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10811674 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10811676 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10811670 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10811669 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10811672 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10811667 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10811675 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10811678 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10811666 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10811671 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10811679 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10811668 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10811663 | - | - |
Affymetrix Rat Clariom S Assay | PSR1900013183.rn.1 | TC1900001638.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1900013185.rn.1 | TC1900001638.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10811663 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21426339 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358966 | - | - |
