Assay Details
Gene Symbol: Mmp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080776.1 | NP_542954.1 | 6 - 7 | 1515 | 86 | |
XM_006237921.2 | XP_006237983.1 | 6 - 7 | 1581 | 86 | ||
GenBank mRNA | D63886.1 | - | 6 - 7 | 1515 | 86 | |
D85509.1 | - | 6 - 7 | 1515 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P552640 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446679 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774688 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534690 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475113 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938012 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89175 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966513 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969196 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63734 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398343 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967156 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535745 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234797 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926229 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347030 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47428 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143369 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345486 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905457 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63196 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340256 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912449 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243383 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823188 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224575 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332744 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48449 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732842 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848189 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939454 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295461 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91301 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556217 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108065 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706813 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747348 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598564 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670012 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81843 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152526 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374083 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913467 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939691 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335584 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382393 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654864 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836715 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910400 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3608 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152606 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291650 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652227 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68726 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586137 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820575 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593934 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883120 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216478 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270562 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605842 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757558 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115279 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327940 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944763 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399936 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938892 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166107 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75184 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656283 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411150 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583508 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306674 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377985 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636208 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946633 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119171 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158188 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157460 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82577 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120662 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142126 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137209 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35900 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12128 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156217 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128344 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74665 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35312 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P552640 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P552640 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P552640 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P142265 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P552640 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701439776 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 78653 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 9811 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 92081 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 35359 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 84081 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 22259 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 40916 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37118 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 59138 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 38524 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 12769 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15419 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 40800 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 25695 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 35531 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 61029 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 73804 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 701439776 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10867767 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867765 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867763 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867772 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867766 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867764 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867771 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867762 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867769 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867770 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867768 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10867761 | - | - |
Rat Genome 230 2.0 Array | 1368590_PM_at | - | - |
Rat Genome 230 2.0 Array | 1368590_at | - | - |
Affymetrix HT Rat Focus Array | 1368590_at | - | - |
Affymetrix HT Rat Focus Array | 1368590_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500001757.rn.1 | TC0500000274.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500001759.rn.1 | TC0500000274.rn.1 | RTA TC |
Rat Expression Set 230 | 1368590_PM_at | - | - |
Rat Expression Set 230 | 1368590_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867761 | - | - |
Rat Genome U34 Set | D63886_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21530083 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361318 | - | - |
RatRef-12 Expression BeadChip | ILMN_70077 | - | - |
