Assay Details
Gene Symbol: Mrps22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077530.1 | NP_001070998.1 | 4 - 5 | 654 | 92 | |
GenBank mRNA | BC105629.1 | - | 4 - 5 | 654 | 7322855 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
