Assay Details
Gene Symbol: Smarcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011966.1 | NP_001011966.1 | 5 - 6 | 555 | 76 | |
XM_006235883.2 | XP_006235945.1 | 4 - 5 | 616 | 76 | ||
XM_006235884.2 | XP_006235946.1 | 4 - 5 | 616 | 76 | ||
XM_006235885.2 | XP_006235947.1 | 4 - 5 | 616 | 76 | ||
XM_006235886.2 | XP_006235948.1 | 5 - 6 | 603 | 76 | ||
XM_006235887.2 | XP_006235949.1 | 5 - 6 | 603 | 76 | ||
XM_006235888.2 | XP_006235950.1 | 4 - 5 | 616 | 76 | ||
XM_006235889.2 | XP_006235951.1 | 4 - 5 | 616 | 76 | ||
GenBank mRNA | BC085349.1 | - | 5 - 6 | 555 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
