Assay Details
Gene Symbol: Gnao1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017327.1 | NP_059023.1 | 1 - 2 | 821 | 72 | |
XM_008772320.1 | XP_008770542.1 | 1 - 2 | 830 | 72 | ||
XM_008772321.1 | XP_008770543.1 | 1 - 2 | 830 | 72 | ||
GenBank mRNA | M17526.1 | - | 1 - 2 | 821 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
