Assay Details
Gene Symbol: Rab14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053589.1 | NP_446041.1 | 3 - 4 | 292 | 69 | |
XM_006234049.2 | XP_006234111.1 | 4 - 5 | 725 | 69 | ||
XM_006234050.2 | XP_006234112.1 | 4 - 5 | 377 | 69 | ||
GenBank mRNA | BC158585.1 | - | 3 - 4 | 881 | 69 | |
M83680.1 | - | 3 - 4 | 292 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
