Assay Details
Gene Symbol: Pah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012619.2 | NP_036751.2 | 9 - 10 | 1056 | 63 | |
XM_008765215.1 | XP_008763437.1 | 7 - 8 | 996 | 63 | ||
GenBank mRNA | BC078881.1 | - | 9 - 10 | 1056 | 63 | |
FQ219397.1 | - | 5 - 6 | 492 | 63 | ||
FQ219680.1 | - | 9 - 10 | 1088 | 63 | ||
K02599.1 | - | 4 - 5 | 346 | 63 | ||
M12337.1 | - | 9 - 10 | 1041 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
