Assay Details
Gene Symbol: Kank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108989.1 | NP_001102459.1 | 9 - 10 | 2333 | 74 | |
XM_008765202.1 | XP_008763424.1 | 9 - 10 | 2390 | 74 | ||
GenBank mRNA | BC166847.1 | - | 9 - 10 | 2316 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
