Assay Details
Gene Symbol: Emc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106496.1 | NP_001099966.1 | 2 - 3 | 235 | 75 | |
XM_006234705.1 | XP_006234767.1 | 2 - 3 | 420 | 75 | ||
GenBank mRNA | FQ212852.1 | - | 2 - 3 | 387 | 75 | |
FQ214203.1 | - | 2 - 3 | 387 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
