Assay Details
Gene Symbol: Efcab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106930.1 | NP_001100400.1 | 3 - 4 | 451 | 88 | |
XM_008768871.1 | XP_008767093.1 | 3 - 4 | 482 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
