Assay Details
Gene Symbol: Pcgf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129882.1 | NP_001123354.1 | 7 - 8 | 880 | 118 | |
XM_006231311.2 | XP_006231373.1 | 7 - 8 | 973 | 118 | ||
XM_008760377.1 | XP_008758599.1 | 6 - 7 | 598 | 118 | ||
GenBank mRNA | BC167067.1 | - | 7 - 8 | 880 | 118 | |
FQ220230.1 | - | 5 - 6 | 471 | 118 | ||
FQ233837.1 | - | 7 - 8 | 815 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
